KEY423214720 Key Organics / BIONET

pentyl 4-hydroxy-3,5-dimethoxybenzoate

  • MDL number MFCD27980619
  • Cas Number : 223907-48-0
  • Molecular Weight : 268.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73 - 74
Purity 97
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431287

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/RS-1032_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart