KEY423214724 Key Organics / BIONET

5-phenyl-2,3-dihydro-1H-1,4-benzodiazepin-2-one

  • MDL number MFCD00277779
  • Cas Number : 2898-08-0
  • Molecular Weight : 236.274

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431291

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4716_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart