KEY424062378 Key Organics / BIONET

3-(2-bromo-5-methoxyphenyl)-2,4-diaza-3-boratricyclo[7.3.1.0^{5,13}]trideca-1(13),5,7,9,11-pentaene

  • MDL number MFCD28016116
  • Molecular Weight : 353.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432055

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2905_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart