KEY424062394 Key Organics / BIONET

(Z)-(1-amino-2-{[(tert-butoxy)carbonyl]amino}ethylidene)amino 5-fluoro-2-(trifluoromethyl)benzoate

  • MDL number MFCD28016112
  • Cas Number : 1610379-67-3
  • Molecular Weight : 379.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432046

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9W-0008_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart