KEY424062407 Key Organics / BIONET

4-bromo-2-(2-methoxyethoxy)benzonitrile

  • MDL number MFCD28015463
  • Cas Number : 1364487-15-9
  • Molecular Weight : 256.099

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432058

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2908_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart