KEY424062410 Key Organics / BIONET

4-bromo-2-[(furan-3-yl)methoxy]benzonitrile

  • MDL number MFCD28015464
  • Cas Number : 1597781-25-3
  • Molecular Weight : 278.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432061

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2911_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart