KEY424062418 Key Organics / BIONET

3-[4-(morpholine-4-carbonyl)phenyl]-2,4-diaza-3-boratricyclo[7.3.1.0^{5,13}]trideca-1(13),5,7,9,11-pentaene

  • MDL number MFCD22124728
  • Molecular Weight : 357.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432069

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2919_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart