KEY424062424 Key Organics / BIONET

(9H-fluoren-9-yl)methyl N-[(2S)-1-(1H-indol-3-yl)-3-oxopropan-2-yl]carbamate

  • MDL number MFCD28016122
  • Cas Number : 246159-95-5
  • Molecular Weight : 410.473

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166
Purity 97
Storage temp. Store under N2 at -18
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432075

Safety Information

signalword Warning
pcodes P411,P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BK-0705_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart