KEY424062429 Key Organics / BIONET

3-(pyrrolidine-1-sulfonyl)-1,2-dihydropyridin-2-one

  • MDL number MFCD23696726
  • Cas Number : 1551867-04-9
  • Molecular Weight : 228.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432079

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3000_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart