KEY424062435 Key Organics / BIONET

1-[3-(pyrazin-2-yl)-1,2,4-oxadiazol-5-yl]cyclohexan-1-amine dihydrochloride

  • MDL number MFCD28016127
  • Cas Number : 1239842-05-7
  • Molecular Weight : 318.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432085

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3006_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart