KEY424062437 Key Organics / BIONET

3-[5-(piperidin-2-yl)-1,2,4-oxadiazol-3-yl]pyridine dihydrochloride

  • MDL number MFCD28016129
  • Cas Number : 1036471-54-1
  • Molecular Weight : 303.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432087

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3008_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart