KEY424062443 Key Organics / BIONET

1-[(2-chloro-4-fluorophenyl)methyl]-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD18788442
  • Cas Number : 1291839-95-6
  • Molecular Weight : 271.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432093

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3014_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart