KEY424062444 Key Organics / BIONET

1-[(4-ethylphenyl)methyl]-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD06623271
  • Cas Number : 824981-40-0
  • Molecular Weight : 247.294

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432094

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3015_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart