KEY424062445 Key Organics / BIONET

1-(2-fluoro-5-methylphenyl)-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD01630405
  • Cas Number : 346446-24-0
  • Molecular Weight : 237.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432095

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3016_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart