KEY424062446 Key Organics / BIONET

5-oxo-1-(5,6,7,8-tetrahydronaphthalen-1-yl)pyrrolidine-3-carboxylic acid

  • MDL number MFCD09814525
  • Cas Number : 913741-97-6
  • Molecular Weight : 259.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432096

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3017_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart