KEY424062448 Key Organics / BIONET

7-[(3-methylphenyl)methyl]-2H,3H,5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyrazine-3,8-dione

  • MDL number MFCD28016134
  • Cas Number : 1610377-17-7
  • Molecular Weight : 258.281

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432098

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3021_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart