KEY424062452 Key Organics / BIONET

1-{[1-(2-fluorophenyl)-1H-1,2,3-triazol-5-yl]methyl}piperidine-4-carboxylic acid

  • MDL number MFCD22390668
  • Cas Number : 1400540-11-5
  • Molecular Weight : 304.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432102

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3025_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart