KEY424062453 Key Organics / BIONET

3-(2-methoxyphenyl)-4H,5H-[1,2,3]triazolo[1,5-a]quinazolin-5-one

  • MDL number MFCD17780270
  • Cas Number : 896373-15-2
  • Molecular Weight : 292.298

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432103

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3027_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart