KEY424062460 Key Organics / BIONET

(Z)-1-[(3-fluorophenyl)methyl]-N'-hydroxy-1H-1,2,3-triazole-4-carboximidamide

  • MDL number MFCD17780207
  • Cas Number : 1255791-10-6
  • Molecular Weight : 235.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432110

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3035_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart