KEY424062472 Key Organics / BIONET

8-fluoro-1H,2H,3H,4H,5H,10H-benzo[b]1,6-naphthyridin-10-one

  • MDL number MFCD21086549
  • Cas Number : 1338652-61-1
  • Molecular Weight : 218.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432122

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3049_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart