KEY424062473 Key Organics / BIONET

4-amino-3-{[(2-chlorophenyl)methyl]carbamoyl}-1,2-thiazole-5-carboxylic acid

  • MDL number MFCD26962707
  • Cas Number : 1113109-08-2
  • Molecular Weight : 311.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432123

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3050_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart