KEY424062476 Key Organics / BIONET

7-fluoro-6-methoxy-1H-indazol-3-amine

  • MDL number MFCD16631265
  • Cas Number : 1279842-87-3
  • Molecular Weight : 181.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432128

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CK-0224_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart