KEY424062496 Key Organics / BIONET

tert-butyl 4-(piperidin-4-yloxy)piperidine-1-carboxylate

  • MDL number MFCD09261176
  • Cas Number : 845305-83-1
  • Molecular Weight : 284.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432149

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4009_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart