KEY424062497 Key Organics / BIONET

(2R)-butan-2-yl methanesulfonate

  • MDL number MFCD01715285
  • Cas Number : 66194-68-1
  • Molecular Weight : 152.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Colourless to Dark Red Liquid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432150

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4010_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart