KEY424062503 Key Organics / BIONET

2-(2,2,2-trifluoroacetyl)-1,2,3,4-tetrahydroisoquinoline-6-carbaldehyde

  • MDL number MFCD09261214
  • Cas Number : 1224587-26-1
  • Molecular Weight : 257.212

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432156

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4018_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart