KEY424062507 Key Organics / BIONET

5-(4-bromophenyl)-3-methyl-1,2,4-oxadiazole

  • MDL number MFCD03069782
  • Cas Number : 71566-07-9
  • Molecular Weight : 239.072

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432160

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4024_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart