KEY424062511 Key Organics / BIONET

7-hydroxy-3,4-dihydro-2H-1-benzopyran-4-one

  • MDL number MFCD08669475
  • Cas Number : 76240-27-2
  • Molecular Weight : 164.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-hydroxy-2,3-dihydro-4H-chromen-4-one
Inchi Code 1S/C9H8O3/c10-6-1-2-7-8(11)3-4-12-9(7)5-6/h1-2,5,10H,3-4H2
Inchi Key XJWBVGDAFGVJEG-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432164

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4029_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart