KEY424062513 Key Organics / BIONET

1-cyclobutylpiperazine dihydrochloride

  • MDL number MFCD00039022
  • Cas Number : 799557-65-6
  • Molecular Weight : 213.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432166

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4033_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart