KEY424062518 Key Organics / BIONET

3-{4-[(tert-butoxy)carbonyl]piperazin-1-yl}benzoic acid

  • MDL number MFCD06657287
  • Cas Number : 193818-13-2
  • Molecular Weight : 306.362

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432171

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4044_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart