KEY424062521 Key Organics / BIONET

6-bromo-2-fluoro-3-methoxybenzoic acid

  • MDL number MFCD09261247
  • Cas Number : 935534-45-5
  • Molecular Weight : 249.035

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432174

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4048_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart