KEY424062531 Key Organics / BIONET

5-chloro-2-methylpyridine-3-carboxylic acid

  • MDL number MFCD11557228
  • Cas Number : 1092286-30-0
  • Molecular Weight : 171.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 207 - 209
Iupac Name 5-chloro-2-methylnicotinic acid
Inchi Code 1S/C7H6ClNO2/c1-4-6(7(10)11)2-5(8)3-9-4/h2-3H,1H3,(H,10,11)
Inchi Key DXQDHJSBBMMJOY-UHFFFAOYSA-N

Description

Melting Point 207 - 209
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432186

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/DK-0705_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart