KEY424062533 Key Organics / BIONET

4-bromo-1-(4-methylbenzenesulfonyl)-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD10575009
  • Cas Number : 348640-07-3
  • Molecular Weight : 351.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432188

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DK-0707_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart