KEY424062535 Key Organics / BIONET

2,6-dimethyl-3-nitroaniline

  • MDL number MFCD00156348
  • Cas Number : 67083-28-7
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,6-dimethyl-3-nitroaniline
Inchi Code 1S/C8H10N2O2/c1-5-3-4-7(10(11)12)6(2)8(5)9/h3-4H,9H2,1-2H3
Inchi Key LDFJDBAOBVUXMQ-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432190

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/DK-0714_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart