KEY424062537 Key Organics / BIONET

4-(8-chloro-1,7-naphthyridin-6-yl)cyclohexane-1-carboxylic acid

  • MDL number MFCD22199656
  • Cas Number : 880466-46-6
  • Molecular Weight : 290.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 217 - 218
Purity 0.97
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432192

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/EH-0706_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart