KEY424062543 Key Organics / BIONET

2-({bicyclo[2.2.1]hept-5-en-2-yl}methyl)-1,1,1,3,3,3-hexafluoropropan-2-ol

  • MDL number MFCD25121854
  • Cas Number : 196314-61-1
  • Molecular Weight : 274.206

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Bpt: >200
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432199

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H314,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-0002_SDS.pdf
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart