KEY424062549 Key Organics / BIONET

(1R)-1-[4-(trifluoromethyl)phenyl]prop-2-en-1-amine hydrochloride

  • MDL number MFCD12068560
  • Cas Number : 1181394-23-9
  • Molecular Weight : 237.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432209

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4767_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart