KEY424062551 Key Organics / BIONET

3-(2-methoxyphenyl)-1-[(4-methylbenzene)sulfonyl]-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD28016157
  • Cas Number : 875639-17-1
  • Molecular Weight : 504.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432211

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4769_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart