KEY424062569 Key Organics / BIONET

(3S,3aS,6R,6aS)-6-{[(4-methylbenzene)sulfonyl]oxy}-hexahydrofuro[3,2-b]furan-3-yl 4-methylbenzene-1-sulfonate

  • MDL number MFCD18428696
  • Cas Number : 66966-12-9
  • Molecular Weight : 454.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432229

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4790_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart