KEY424062575 Key Organics / BIONET

ethyl 2-acetamido-7-oxo-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

  • MDL number MFCD00437065
  • Cas Number : 91421-24-8
  • Molecular Weight : 281.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432235

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4796_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart