KEY424062580 Key Organics / BIONET

4-(benzyloxy)cyclohexan-1-one

  • MDL number MFCD10697867
  • Molecular Weight : 204.269

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/432240

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4801_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart