KEY424826496 Key Organics / BIONET

6-methoxy-2-(4-nitrophenyl)-1,3-benzothiazole

  • MDL number MFCD06410799
  • Cas Number : 43036-14-2
  • Molecular Weight : 286.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433304

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11X-0248_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart