KEY424826506 Key Organics / BIONET

methyl pyrimidine-4-carboxylate

  • MDL number MFCD00234184
  • Cas Number : 2450-08-0
  • Molecular Weight : 138.126

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70 - 72
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433318

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9N-070_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart