KEY424826507 Key Organics / BIONET

methyl 2,6-dioxo-3-{[2-(trimethylsilyl)ethoxy]methyl}-1,2,3,6-tetrahydropyrimidine-4-carboxylate

  • MDL number MFCD28023508
  • Cas Number : 1610377-18-8
  • Molecular Weight : 300.386

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433319

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9Y-0023_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart