KEY424826508 Key Organics / BIONET

7-methyl-6-nitro-1H-indazole

  • MDL number MFCD16620021
  • Cas Number : 208457-81-2
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 221 - 225
Iupac Name 7-methyl-6-nitro-1H-indazole
Inchi Code 1S/C8H7N3O2/c1-5-7(11(12)13)3-2-6-4-9-10-8(5)6/h2-4H,1H3,(H,9,10)
Inchi Key YKHCFVZVZBXQBH-UHFFFAOYSA-N

Description

Melting Point 221 - 225
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433320

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/AH-0082_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart