KEY424826515 Key Organics / BIONET

8-methyl-3-phenyl-1,4,8-triazaspiro[4.5]dec-3-ene-2-thione

  • MDL number MFCD14726872
  • Cas Number : 892299-50-2
  • Molecular Weight : 259.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433326

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3053_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart