KEY424826517 Key Organics / BIONET

2-(2H-1,3-benzodioxol-5-yl)-4H,5H-pyrazolo[1,5-a]pyrazin-4-one

  • MDL number MFCD17780274
  • Cas Number : 1255777-72-0
  • Molecular Weight : 255.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433328

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3055_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart