KEY424826519 Key Organics / BIONET

1-(3-cyanoquinolin-4-yl)piperidine-4-carboxylic acid

  • MDL number MFCD13762738
  • Cas Number : 1156272-06-8
  • Molecular Weight : 281.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433330

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3057_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart