KEY424826524 Key Organics / BIONET

1-(3-phenyl-1H-pyrazole-5-carbonyl)piperidin-4-amine

  • MDL number MFCD20528921
  • Cas Number : 1607174-15-1
  • Molecular Weight : 270.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433335

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3063_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart