KEY424826527 Key Organics / BIONET

1-ethyl-1H,6H,7H-pyrrolo[2,3-c]pyridin-7-one

  • MDL number MFCD20441705
  • Cas Number : 1340902-81-9
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433338

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3066_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart