KEY424826529 Key Organics / BIONET

1-(2-methoxyethyl)-1H,6H,7H-pyrrolo[2,3-c]pyridin-7-one

  • MDL number MFCD20528978
  • Cas Number : 1340857-27-3
  • Molecular Weight : 192.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433340

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3068_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart